
2-(Diphenylphosphino)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50777-76-9
:
Formula:C₁₉H₁₅OP
MDL Number:MFCD00013367
Melting Pt:112…115 °C
:290.30 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together