Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Catalog # APOSOR322194-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Catalog # APOSOR322194-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :198.68 g/mol
Storage Temperature:Ambient
CAS Number:108831-68-1
Formula:C₈H₇ClN₂S
MDL Number:MFCD00464832