Order Entry
Czech Republic
ContactUsLinkComponent
3-Hydroxyphthalic anhydride 97%
Irritant
3-Hydroxyphthalic anhydride 97%
Catalog # APOSOR322192-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Hydroxyphthalic anhydride 97%
Catalog # APOSOR322192-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :164.1150 g/mol
Melting Pt:199…202 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:37418-88-5
Formula:C₈H₄O₄
MDL Number:MFCD00011557