Order Entry
Czech Republic
ContactUsLinkComponent
 
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Catalog # APOSOR322169-5G
Supplier:  Apollo Scientific
CAS Number:  
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Catalog # APOSOR322169-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :335.55 g/mol
Storage Temperature:Ambient
CAS Number:14933-08-5
Formula:C₁₇H₃₇NO₃S
MDL Number:MFCD00036909