

p-Toluenesulphonic acid monohydrate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6192-52-5
:
Formula:C₇H₈O₃S·1H₂O
MDL Number:MFCD00142137
:190.22 g/mol
Boiling Pt:140 °C/20mmHg
Melting Pt:103…106 °C
Density:1.24 g/cm³ (25 °C)
Flash Pt:150 °C
Storage Temperature:Ambient
UN:2585
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together