Order Entry
Czech Republic
ContactUsLinkComponent
2-(3,4-Dichlorophenyl)ethanol
2-(3,4-Dichlorophenyl)ethanol
Catalog # APOSOR322080-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3,4-Dichlorophenyl)ethanol
Catalog # APOSOR322080-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :191.05 g/mol
Storage Temperature:Ambient
CAS Number:35364-79-5
Formula:C₈H₈Cl₂O
MDL Number:MFCD00800673