Order Entry
Czech Republic
ContactUsLinkComponent
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
Catalog # APOSOR32187-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
Catalog # APOSOR32187-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.69 g/mol
Storage Temperature:Ambient
CAS Number:109925-12-4
Formula:C₁₂H₁₁ClN₂O₂
MDL Number:MFCD00215393