Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Chlorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid, Technical Grade
2-(4-Chlorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid, Technical Grade
Catalog # APOSOR32172-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Chlorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid, Technical Grade
Catalog # APOSOR32172-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.72 g/mol
Storage Temperature:Ambient
CAS Number:78364-19-9
Formula:C₁₆H₁₀ClNO₃
MDL Number:MFCD00140321