
1H-Pyrido[2,3-d][1,3]oxazine-2,4-dione 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21038-63-1
:
Formula:C₇H₄N₂O₃
MDL Number:MFCD09863994
:164.12 g/mol
Melting Pt:207…208 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together