Order Entry
Czech Republic
ContactUsLinkComponent
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride 97%
Irritant
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride 97%
Catalog # APOSOR321313-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride 97%
Catalog # APOSOR321313-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.6339 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:666261-01-4
Formula:C₆H₁₄ClNO₂
MDL Number:MFCD18072019