Order Entry
Czech Republic
ContactUsLinkComponent
1,1-Diethoxy-2-nitroethane 95%
Irritant
1,1-Diethoxy-2-nitroethane 95%
Catalog # APOSOR321286-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1-Diethoxy-2-nitroethane 95%
Catalog # APOSOR321286-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :163.17
Boiling Pt:89…91 °C/14mmHg
 :GHS07
Storage Temperature:Refrigerator
CAS Number:34560-16-2
Formula:C₆H₁₃NO₄