3-Hydroxy-6-methyl-2,3-dihydro-4H-furo[3,2-c]pyran-4-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:57053-18-6
:
Formula:C₈H₈O₄
MDL Number:MFCD01443822
:168.15 g/mol
Storage Temperature:Ambient
Frequently Bought Together