3-(Dimethylamino)-2-(3-thienyl)acrylonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (2Z)-3-(Dimethylamino)-2-(thiophen-3-yl)prop-2-enenitrile
CAS Number:165404-39-7
:
Formula:C₉H₁₀N₂S
MDL Number:MFCD00127051
Molecular Weight:178.26 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together