Methyl-2-amino-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6497-03-6
:
Formula:C₈H₁₁ClN₂O₄S
MDL Number:MFCD11052312
:266.7 g/mol
Storage Temperature:Ambient
Frequently Bought Together