Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-2-Aminomethyl-1-boc-azetidine
(S)-2-Aminomethyl-1-boc-azetidine
(S)-2-Aminomethyl-1-boc-azetidine
(S)-2-Aminomethyl-1-boc-azetidine
APOSOR317248-1G
:
Apollo Scientific
:
(S)-2-Aminomethyl-1-boc-azetidine
APOSOR317248-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
186.26 g/mol
:
GHS07
:
Refrigerator
:
1007873-90-6
:
C₉H₁₈N₂O₂
:
MFCD17215571