
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) 97%
Catalog # APOSOR317237-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.15 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1045709-32-7
:MFCD16652332
Formula:C₁₂H₂₀N₂O₆