3,3-Dimethoxycyclobutane-1-carboxylate methyl ester 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98231-07-3
:
Formula:C₈H₁₄O₄
MDL Number:MFCD11111944
:174.2 g/mol
Storage Temperature:Ambient
Frequently Bought Together