Order Entry
Czech Republic
ContactUsLinkComponent
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Catalog # APOSOR316007-100G
Supplier:  Apollo Scientific
CAS Number:  
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Catalog # APOSOR316007-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :245.28 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:102195-79-9
Formula:C₁₁H₁₉NO₅
MDL Number:MFCD00237541