Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Chlorophenoxy)aniline 98%
2-(4-Chlorophenoxy)aniline 98%
Catalog # APOSOR315743-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Chlorophenoxy)aniline 98%
Catalog # APOSOR315743-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :219.6700 g/mol
Storage Temperature:Ambient
CAS Number:2770-11-8
Formula:C₁₂H₁₀ClNO
MDL Number:MFCD00025168