
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 95+%
Katalogové číslo APOSOR315579-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:236.3100 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:108468-00-4
Formula:C₁₃H₂₀N₂O₂
MDL Number:MFCD02683058