Order Entry
Czech Republic
ContactUsLinkComponent
(S)-3-Methylmorpholine hydrochloride 97%
(S)-3-Methylmorpholine hydrochloride 97%
Catalog # APOSOR315414-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-3-Methylmorpholine hydrochloride 97%
Catalog # APOSOR315414-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :137.61 g/mol
Storage Temperature:Ambient
CAS Number:1022094-03-6
Formula:C₅H₁₁NO·ClH
MDL Number:MFCD18382512