
Diethyl-trans-1,2-cyclopropanedicarboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3999-55-1
:
Formula:C₉H₁₄O₄
MDL Number:MFCD00198002
:186.207 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together