Order Entry
Czech Republic
ContactUsLinkComponent
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose
Catalog # APOSOR315347-25G
Supplier:  Apollo Scientific
CAS Number:  
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose
Catalog # APOSOR315347-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :260.29 g/mol
Storage Temperature:Ambient
CAS Number:20880-92-6
Formula:C₁₂H₂₀O₆
MDL Number:MFCD00022183