Order Entry
Czech Republic
ContactUsLinkComponent
6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride 97+%
6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride 97+%
Catalog # APOSOR315310-100MG
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride 97+%
Catalog # APOSOR315310-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :170.64 g/mol
Storage Temperature:Ambient
CAS Number:1187930-42-2
Formula:C₈H₁₁ClN₂
MDL Number:MFCD12913793