
Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:37031-29-1
:
Formula:C₉H₁₄O₆
MDL Number:MFCD00066218
:218.21 g/mol
Boiling Pt:150 °C/19mmHg
Density:1.19 g/cm³ (25 °C)
Flash Pt:110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together