Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dichlorophenyl isothiocyanate
2,6-Dichlorophenyl isothiocyanate
Catalog # APOSOR315263-5G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichlorophenyl isothiocyanate
Catalog # APOSOR315263-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :204.08 g/mol
Storage Temperature:Ambient
CAS Number:6590-95-0
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00041056