Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxy-5-methylphenyl isothiocyanate
2-Methoxy-5-methylphenyl isothiocyanate 99%
2-Methoxy-5-methylphenyl isothiocyanate 99%
APOSOR315241-1G
:
Apollo Scientific
:
2-Methoxy-5-methylphenyl isothiocyanate 99%
APOSOR315241-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
179.24 g/mol
:
Ambient
:
190774-56-2
:
C₉H₉NOS
:
MFCD00041083