Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-5-iodo-4-methylpyridine 97%
2-Bromo-5-iodo-4-methylpyridine 97%
Catalog # APOSOR315201-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-5-iodo-4-methylpyridine 97%
Catalog # APOSOR315201-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :297.92 g/mol
Storage Temperature:Ambient
CAS Number:942206-07-7
Formula:C₆H₅BrIN
MDL Number:MFCD09037465