Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52373-72-5
:
Formula:CâHââOâ
MDL Number:MFCD00066527
:160.17 g/mol
Storage Temperature:Ambient
Frequently Bought Together