
3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
Catalog # APOSOR3136-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:216.29 g/mol
Melting Pt:108…113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:6631-89-6
Formula:C₁₃H₁₆N₂O
MDL Number:MFCD00051762