Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4-(bromomethyl)-3-chlorobenzoate
Methyl-4-(bromomethyl)-3-chlorobenzoate
Methyl-4-(bromomethyl)-3-chlorobenzoate
APOSOR313095-5G
:
Apollo Scientific
:
Methyl-4-(bromomethyl)-3-chlorobenzoate
APOSOR313095-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
263.52 g/mol
:
Ambient
:
74733-30-5
:
C₉H₈BrClO₂
:
MFCD10566565