
1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol
Catalog # APOSOR313043-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:334.3600 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:10535-17-8
Formula:C₁₈H₂₂O₆
MDL Number:MFCD01880862