2-Oxa-6-azaspiro[3.4]octan-7-one
Catalog # APOSOR312208-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:127.14 g/mol
Storage Temperature:Ambient
CAS Number:1207174-87-5
Formula:C₆H₉NO₂
MDL Number:MFCD14586414