Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Tosylazetidin-3-one
1-Tosylazetidin-3-one
1-Tosylazetidin-3-one
1-Tosylazetidin-3-one
APOSOR312042-250MG
:
Apollo Scientific
:
1-Tosylazetidin-3-one
APOSOR312042-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
225.27 g/mol
:
Ambient
:
76543-27-6
:
C₁₀H₁₁NO₃S
:
MFCD00129860