Order Entry
Czech Republic
ContactUsLinkComponent
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one
Catalog # APOSOR310711-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one
Catalog # APOSOR310711-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :338.39 g/mol
Storage Temperature:Ambient
CAS Number:892300-30-0
Formula:C₁₈H₁₄N₂O₃S
MDL Number:MFCD07655393