2-(4-Methoxyphenyl)-4-oxo-4,5-dihydropyrazolo[1,5-a]pyrazine-3-carbaldehyde
Catalog # APOSOR310700-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.26 g/mol
Storage Temperature:Ambient
CAS Number:1610377-12-2
Formula:C₁₄H₁₁N₃O₃
MDL Number:MFCD28023509