Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
L-Prolinamide 97%

L-Prolinamide 97%

Irritant
Supplier:   Apollo Scientific
L-Prolinamide 97%
Supplier:  Apollo Scientific
Certificates
L-Prolinamide 97%

2 Options Available Below

Product Details & Documents
Synonyms:  H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2
  • CAS Number:7531-52-4
  • Formula:C₅H₁₀N₂O
  • MDL Number:MFCD00005253
  • Molecular Weight:114.15 g/mol
  • Melting Pt:95…97 °C
  • Storage Temperature:Ambient
  • EINECS:231-397-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 440,00 Kč
    1 440,00 Kč
    1 * 5 g
       1 440,00 Kč

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 850,00 Kč
    1 850,00 Kč
    1 * 25 g
       1 850,00 Kč

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.