6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole
Catalog # APOSOR310656-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.31 g/mol
Storage Temperature:Ambient
CAS Number:43036-14-2
Formula:C₁₄H₁₀N₂O₃S
MDL Number:MFCD06410799