Ethyl-2-cyclohexanoneacetate
Catalog # APOSOR310644-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:184.24 g/mol
Storage Temperature:Ambient
CAS Number:24731-17-7
Formula:C₁₀H₁₆O₃
MDL Number:MFCD00001638