Order Entry
Czech Republic
ContactUsLinkComponent
Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
Catalog # APOSOR310605-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
Catalog # APOSOR310605-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.1406 g/mol
Storage Temperature:Ambient
CAS Number:64700-65-8
Formula:C₆H₉NO₃
MDL Number:MFCD01318621