1-Butyn-3-yl-p-toluenesulphonate
Catalog # APOSOR310300-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:224.27 g/mol
Storage Temperature:Ambient
CAS Number:53487-52-8
Formula:C₁₁H₁₂O₃S
MDL Number:MFCD00060095