Order Entry
Czech Republic
ContactUsLinkComponent
(1R,2S,5S)-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥99%, ee 99%
Irritant
(1R,2S,5S)-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥99%, ee 99%
Catalog # APOSOR310264-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(1R,2S,5S)-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥99%, ee 99%
Catalog # APOSOR310264-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :127.14 g/mol
Melting Pt:252…256 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:33294-81-4
Formula:C₆H₉NO₂
MDL Number:MFCD05663832