Order Entry
Czech Republic
ContactUsLinkComponent
 
2,2'-Dichloro-4,4'-bipyridyl
Catalog # APOSOR310158-100MG
Supplier:  Apollo Scientific
CAS Number:  
2,2'-Dichloro-4,4'-bipyridyl
Catalog # APOSOR310158-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :225.08 g/mol
Storage Temperature:Ambient
CAS Number:53344-74-4
Formula:C₁₀H₆Cl₂N₂
MDL Number:MFCD01106345