
4-Chloro-1,6-naphthyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6861-84-3
:
Formula:C₈H₅ClN₂
MDL Number:MFCD09881194
:164.6 g/mol
Melting Pt:90 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together