Order Entry
Czech Republic
ContactUsLinkComponent
Pyrido[3,4-d]pyrimidin-4(3H)-one
Irritant
Pyrido[3,4-d]pyrimidin-4(3H)-one
Catalog # APOSOR310008-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Pyrido[3,4-d]pyrimidin-4(3H)-one
Catalog # APOSOR310008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:320 °C (dec.)
 :147.1300 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:19178-25-7
Formula:C7H5N3O
MDL Number:MFCD09999183