Order Entry
Czech Republic
ContactUsLinkComponent
6-Bromoimidazo[1,2-a]pyridin-3-amine
Irritant
6-Bromoimidazo[1,2-a]pyridin-3-amine
Catalog # APOSOR30980-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromoimidazo[1,2-a]pyridin-3-amine
Catalog # APOSOR30980-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :212.05 g/mol
Melting Pt:145…147 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:705262-55-1
Formula:C₇H₆BrN₃
MDL Number:MFCD09864901