
2-Methylimidazo[1,2-a]pyridin-3-amine 95%
Catalog # APOSOR30979-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:147.18 g/mol
Melting Pt:134…136 °C
:GHS07
Storage Temperature:Ambient
CAS Number:28036-31-9
Formula:C₈H₉N₃
MDL Number:MFCD09800571