Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Irritant
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Catalog # APOSOR30958-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Catalog # APOSOR30958-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :295.39 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1171901-11-3
Formula:C₁₄H₂₅N₅O₂
MDL Number:MFCD11505005