
4-(2-Chlorophenyl)-1,3-thiazol-2-amine
Catalog # APOSOR30955-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:210.69 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:21344-90-1
Formula:C₉H₇ClN₂S
MDL Number:MFCD01315352