Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Diethyl-4-nitrosoaniline 98%
N,N-Diethyl-4-nitrosoaniline 98%
Catalog # APOSOR30878-25G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Diethyl-4-nitrosoaniline 98%
Catalog # APOSOR30878-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :178.23 g/mol
Storage Temperature:Ambient
CAS Number:120-22-9
Formula:C₁₀H₁₄N₂O
MDL Number:MFCD00002064